Skip to Content
Merck
All Photos(1)

Key Documents

A4479

Sigma-Aldrich

Anti-α1B Adrenergic Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 3 μg/mL using artery, Tunica intima and Tunica media

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRA1B(147)

General description

Adrenergic receptors (ARs) belong to the G protein-coupled receptor superfamily that interacts with epinephrine and norepinephrine. There are nine receptor subtypes which include three α1AR subtypes (α1A/D, α1B, and α1C), three α2ARs (α2A, α2B, and α2C), and three βAR subtypes (β1, β2, and β3). Adrenergic receptors have been implicated in many diseases such as hypertension, glaucoma, cardiac disorders, diabetes, impotence and depression. Rabbit Anti-α1B Adrenergic Receptor antibody specifically recognizes human α1B adrenergic receptor.

Immunogen

synthetic peptide corresponding to the C-terminal domain of human α1B adrenergic receptor. The immunizing peptide has 85% homology with the rat gene and 90% homology with the mouse gene.

Application

Anti-α1B Adrenergic Receptor antibody can be used for immunohistochemistry at 3μg/ml using artery, tunica intima and tunica media tissues.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service