Skip to Content
Merck

Skip To

A4086

Anti-ATF-2 (SS-16) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, IP, WB CL
Citations:
5


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 65 kDa

species reactivity

human

technique(s)

immunoprecipitation (IP): 5 μg using lysates of human K562 cells, microarray: suitable, western blot (chemiluminescent): 1:250 using whole cell extract of human K562 or Jurkat cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATF2(1386)

General description

ATF2 (Activating Transcription Factor 2, CRE-BP,HB-16, CREB2, TREB-7) belongs to the ATF/CREB family of basic region leucine zipper DNAbinding proteins. ATF2 regulates transcription by binding to a consensus cAMP response element (CRE) in the promoter of various viral and cellular genes. Rabbit anti-ATF-2 (SS-16) antibody recognizes human ATF2 by immunoblotting (65kDa) and immunoprecipitation. Staining of ATF2 by immunoblotting is inhibited by the immunizing peptide.

Immunogen

synthetic peptide corresponding to amino acids 490-505 of human ATF-2.

Application

Rabbit anti-ATF-2 can be used for immunoprecipitation (5μg, using lysates of human K562 cells), microarray, and chemiluminescent western blot (1:250, using whole cell extract of human K562 or Jurkat cells).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters