Skip to Content
Merck
All Photos(1)

Key Documents

A3830

Sigma-Aldrich

Adrenomedullin rat

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$1,330.00

$1,330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
$1,330.00

About This Item

Empirical Formula (Hill Notation):
C242H379N77O75S5
CAS Number:
Molecular Weight:
5727.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$1,330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Adm(25026)

Amino Acid Sequence

Tyr-Arg-Gln-Ser-Met-Asn-Gln-Gly-Ser-Arg-Ser-Thr-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Met-Gln-Lys-leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Gly-Met-Ala-Pro-Arg-Asn-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2

Biochem/physiol Actions

Elicits a potent and long-lasting hypotensive activity in anesthetized rats. Plays a key role in regulating endometrial angiogenesis.
Elicits a potent and long-lasting hypotensive effect; plays a key role in regulating endometrial angiogenesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Kesik et al.
Clinical and experimental dermatology, 35(8), 897-901 (2010-03-30)
Extravasation of vesicant drugs such as vinca alkaloids causes severe injury, which may range from erythema to skin necrosis or ulceration. The skin necrosis may not be fully evident until several weeks or months after the initial damage, and may

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service