Skip to Content
Merck

Skip To

A3608

Bovine Serum Albumin

lyophilized powder, ≥96% (agarose gel electrophoresis)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Check Cart for Availability
$463.00
500 g
Check Cart for Availability
$1,610.00

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.21
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥96% (agarose gel electrophoresis)
Form:
lyophilized powder
Technique(s):
immunoprecipitation (IP): suitable

$463.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

assay

≥96% (agarose gel electrophoresis)

form

lyophilized powder

purified by

heat shock fractionation

packaging

poly bottle of

origin

New Zealand origin

technique(s)

immunoprecipitation (IP): suitable

pH

7, ~7 (7 % (w/v) in H2O)

solubility

water: slightly soluble

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

Bovine serum albumin (BSA) is a 66 kDa protein consisting of three domains with two subdomains under each. It is a α-helical, globular and non-glycosylated protein. It belongs to the serum albumins family. It has 17-disulfide bonds. Albumin is the most abundant plasma protein in humans.

Application

Bovine Serum Albumin has been used:
  • as a component in chromatin immunoprecipitation (ChIP IP) buffer
  • in combination with electrospun polystyrene (ESPS) fiber mats for fluorescence microscopy
  • as a component of blocking solution and immobilzation studies
  • as a component in phosphate buffer saline (PBS)

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.
Bovine Serum Albumin (BSA) is a transporter for drugs, hormones and fatty acids. High levels of albumin is associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia.

Preparation Note

Derived from New Zealand source serum
Prepared using heat shock fractionation
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2153A3983A9418
origin

New Zealand origin

origin

USA origin

origin

New Zealand origin

origin

USA origin

technique(s)

immunoprecipitation (IP): suitable

technique(s)

ELISA: suitable, blood typing: suitable, microbiological culture: suitable

technique(s)

western blot: suitable

technique(s)

cell culture | mammalian: suitable

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

assay

≥96% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

assay

≥98% (agarose gel electrophoresis)

assay

≥96% (agarose gel electrophoresis)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters