Skip to Content
Merck
All Photos(2)

Key Documents

A3282

Sigma-Aldrich

5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide ester

≥95%, powder

Synonym(s):

N-(5-Azido-2-nitrobenzoyloxy)succinimide, N-Succinimidyl 5-azido-2-nitrobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7N5O6
CAS Number:
Molecular Weight:
305.20
Beilstein:
1555224
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

Assay

≥95%

form

powder

reaction suitability

reagent type: cross-linking reagent

color

yellow to yellow-orange

solubility

ethyl acetate: 25 mg/mL
DMF: soluble

functional group

NHS ester

storage temp.

2-8°C

SMILES string

O=C(ON1C(CCC1=O)=O)C2=C(C=CC(N=[N+]=[N-])=C2)[N+]([O-])=O

InChI

1S/C11H7N5O6/c12-14-13-6-1-2-8(16(20)21)7(5-6)11(19)22-15-9(17)3-4-10(15)18/h1-2,5H,3-4H2

InChI key

FUOJEDZPVVDXHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Photoactive, heterobifunctional cross-linking reagent. Typically, initial reaction couples via ester to primary amine by amide bond formation in the pH range 6.5-8.5. Second bonding occurs during UV irradiation (250-350 nm) via reactive nitrene. The latter bonding is rapid and non-specific.

Caution

Reducing agents such as thiols may reduce the azide to amine and should be avoided. Initial manipulations and coupling should be performed under reduced light.

Other Notes

Note that the nitro substituent provides an absorption band at a longer wavelength compared to simple aryl azide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Green et al.
FEBS letters, 206(1), 130-134 (1986-09-29)
Rat adipocyte plasma membranes were incubated with the A1 adenosine receptor agonist, 125I-hydroxyphenylisopropyl adenosine (1 nM) and then treated with the photoactive cross-linking agent, ANB-NOS. The membranes were solubilized and analyzed by SDS-PAGE and autoradiography. A single protein, Mr approx.
S P Sheikh et al.
The Journal of biological chemistry, 265(14), 8304-8310 (1990-05-15)
Pharmacological studies indicate that peptide YY (PYY) and neuropeptide Y interact with multiple binding sites, categorized as Y1 and Y2 subtypes. In order to identify and structurally characterize the Y1 and Y2 receptors we covalently cross-linked [125I-Tyr36]PYY to its receptors.
S Karrasch et al.
Biophysical journal, 65(6), 2437-2446 (1993-12-01)
Scanning force microscopy allows imaging of biological molecules in their native state in buffer solution. To this end samples have to be fixed to a flat solid support so that they cannot be displaced by the scanning tip. Here we
P R Buckland et al.
The Biochemical journal, 235(3), 879-882 (1986-05-01)
We have recently shown that the beta subunit of thyrotropin (TSH) can be cross-linked to the TSH receptor [Buckland, Strickland, Pierce & Rees Smith (1985) Endocrinology (Baltimore) 116, 2122-2124; Buckland, Strickland & Rees Smith (1985) Biochem. Soc. Trans. 13, 942-943].
S A McMahan et al.
Biochemistry, 33(40), 12092-12099 (1994-10-11)
In an effort to better understand protein-protein photoaffinity cross-linking using aryl azides, we have tested a number of factors influencing the cross-linking of the sigma 70 subunit of Escherichia coli RNA polymerase to core RNA polymerase. These factors include the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service