Skip to Content
Merck
All Photos(1)

Key Documents

A3171

Sigma-Aldrich

Alternariol monomethyl ether from Alternaria alternata (tenuis)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$703.00

$703.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$703.00

About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.77

$703.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

COc1cc(O)c2C(=O)Oc3cc(O)cc(C)c3-c2c1

InChI

1S/C15H12O5/c1-7-3-8(16)4-12-13(7)10-5-9(19-2)6-11(17)14(10)15(18)20-12/h3-6,16-17H,1-2H3

InChI key

LCSDQFNUYFTXMT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Zhao et al.
Journal of agricultural and food chemistry, 63(1), 343-348 (2014-12-19)
A total of 70 tomato-based and 86 citrus-based products collected in China were analyzed for alternariol, alternariol monomethyl ether, tentoxin, and tenuazonic acid by ultraperformance liquid chromatography–electrospray ionization–tandem mass spectrometry. No toxins were found in any fresh tomato or citrus
Shufang Wang et al.
Journal of pharmaceutical and biomedical analysis, 98, 201-209 (2014-06-18)
This work reported an efficient and accurate liquid chromatography tandem mass spectrometry (LC-MS/MS) method for simultaneous determination of seventeen mycotoxins in Puerariae lobatae radix, a frequently used traditional Chinese medicine (TCM). The effects of four different clean-up methods, including TC-M160

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service