Skip to Content
Merck
All Photos(1)

Key Documents

A271

Sigma-Aldrich

Anti-α2A Adrenergic Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ADRA2, Anti-ADRA2R, Anti-ADRAR, Anti-ALPHA2AAR, Anti-ZNF32

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC (p)
WB
clone:
polyclonal
species reactivity:
rat, human, mouse, pig
citations:
6
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1000
western blot: 1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

rat, human, mouse, pig

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1000
western blot: 1:500

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADRA2A(150)
mouse ... Adra2a(11551)
rat ... Adra2a(25083)

General description

Adrenergic receptors (ARs) belong to the 7-transmembrane domain G protein-coupled receptor superfamily that binds to the endogenous catecholamines, epinephrine and norepinephrine. α2AR subtypes block adenylyl cyclase, suppress voltage-sensitive Ca2+ channels, and stimulate receptor dependent K+ channels. All of the α2AR subtypes inhibit adenylyl cyclase through coupling to members of the Gi/Go class of G proteins. They are an important component of the neural circuitry and modulate cardiovascular physiology. The physiological function of the α2ARs in the kidney is to regulate Na+/H+ exchange.
Anti-α2A adrenergic receptor detects rat and mouse α2A adrenergic receptors by immunoblotting and immunohistochemistry. In immunoblotting, the antibody detects an approx. 45kDa band representing α2A receptors in tissues expressing this receptor subtype, although some non-specific bands have also been observed in certain cases.

Immunogen

synthetic peptide derived from amino acids 218-235 of human, mouse, rat, and pig α2A adrenergic receptor. The immunogen sequence is found within the third intracellular loop of the α2A adrenergic receptor subtype.

Application

Rabbit anti-α2A adrenergic receptor antibody can be used for western blot (1:500) and immunohistochemistry (1:1000) applications.

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elvira Ganic et al.
Cell reports, 14(8), 1991-2002 (2016-02-26)
Monoamine and acetylcholine neurotransmitters from the autonomic nervous system (ANS) regulate insulin secretion in pancreatic islets. The molecular mechanisms controlling neurotransmitter signaling in islet β cells and their impact on diabetes development are only partially understood. Using a glucose-intolerant, MafA-deficient
Wenli Mi et al.
Anesthesia and analgesia, 125(3), 1032-1041 (2017-05-26)
Combination drug therapy is commonly used to treat chronic pain conditions such as neuropathic pain, and antidepressant is often used together with opioid analgesics. While rewarding is an intrinsic property of opioid analgesics, it is unknown whether the use of
Olena Fedotkina et al.
Frontiers in neuroscience, 16, 858049-858049 (2022-05-24)
Persons with type 2 diabetes born in the regions of famine exposures have disproportionally elevated risk of vision-threatening proliferative diabetic retinopathy (PDR) in adulthood. However, the underlying mechanisms are not known. In the present study, we aimed to investigate the
Shigekazu Sugino et al.
Genes, brain, and behavior, e12759-e12759 (2021-06-12)
The genetic mechanisms of postoperative nausea and vomiting (PONV) and the involvement of the catecholamine system in the brain have not been elucidated. Eating kaolin clay as a type of pica has been examined as an alternative behavior to emesis.
Dong Ji et al.
Experimental and therapeutic medicine, 14(5), 4910-4918 (2017-12-05)
As a therapeutic target for neuropathic pain, the anti-nociceptive effects of α 2-adrenoceptors (α2AR) have attracted attention. Dexmedetomidine (DEX), a potent and highly selective α2AR agonist, has exhibited significant analgesic effects in neuropathic pain, but the underlying mechanism has remained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service