Skip to Content
Merck
All Photos(1)

Key Documents

A2668

Sigma-Aldrich

Anti-Actin antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

mouse, rat, human, chicken

technique(s)

indirect immunofluorescence: 1:40 using cultured chicken fibroblasts
western blot: suitable using rabbit skeletal muscle α-actin1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ACTA1(58)
mouse ... Acta1(11459)
rat ... Acta1(29437)

General description

Actin, a highly conserved protein, is a major component of both the cytoskeletal and contractile structures in all cell types. It varies in amount, being related to the type of differentiation and to the func tional state of cells and tissues. Actin can be found in two different forms of aggregation, the globular or the fibrillar state, and at least six distinct isoforms occur in vertebrates. The actins exhibit over 90% sequence homology, but each isoform has a unique NH2-terminal sequence. The isoforms are comprised of three α actins (skeletal, cardiac, smooth), one β actin (β-non-muscle) and two γ actins (γ smooth muscle and γ non-muscle). Difficulties have been encountered in producing polyclonal antibodies due to the highly conserved nature of actin.
Rabbit Anti-Actin reacts with pure actin by counter immunoelectrophoresis. The product also stains the I-bands of stretched skeletal muscle.
Rabbit polyclonal anti-Actin antibody reacts with rat, chicken, human, and mouse actin. By immunoblotting, cross-reactivity with other proteins may be observed.

Immunogen

actin from chicken back muscle

Application

May be used for studying the intracellular distribution of actin structures and their functions in the contractile system.
Rabbit anti-actin antibody has been used for western blot assays using mouse lung homogenates and human umbilical artery endothelial cell (HUAEC) lysates.
Rabbit anti-actin antibody was used to visualize actin in human sperm smears. Samples were incubated with the antibody (1:80) for 2 hours at 37 degrees.
Fluorescent microscopy was performed on bovine fallopian tube epithilial cells to visualize actin aggregates following inoculation with c. parvum sporozites using the rabbit anti-actin antibody from Sigma.
Rabbit polyclonal anti-Actin antibody is used to tag actin for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of actin in stress fibers and I-bands of skeletal muscle cells.

Physical form

Rabbit Anti-Actin is supplied as a liquid containing 0.1% sodium azide as preservative.

Other Notes

To view an Actin antibody selection guide, please visit www.sigmaaldrich.com/actin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Bouvier et al.
American journal of physiology. Renal physiology, 296(1), F160-F169 (2008-11-07)
Calcineurin inhibitors cyclosporine and tacrolimus are effective immunosuppressants, but both substances have the same intrinsic nephrotoxic potential that adversely affects allograft survival in renal transplant patients and causes end-stage renal disease in other solid organ or bone marrow transplant recipients.
Jianfei Jiang et al.
Radiation research, 172(4), 414-422 (2009-09-24)
To assist in screening existing drugs for use as potential radioprotectors, we used a human unbiased 16,560 short interfering RNA (siRNA) library targeting the druggable genome. We performed a synthetic protection screen that was designed to identify genes that, when
Ciric To et al.
The Journal of biological chemistry, 285(36), 27944-27957 (2010-06-23)
Synthetic triterpenoids are anti-tumor agents that affect numerous cellular functions including apoptosis and growth inhibition. Here, we used mass spectrometric and protein array approaches and uncovered that triterpenoids associate with proteins of the actin cytoskeleton, including actin-related protein 3 (Arp3).
Farah Ketroussi et al.
PloS one, 6(8), e22776-e22776 (2011-09-03)
Human leukocyte antigen G (HLA-G) is involved in regulating T-cell responses through its interaction with inhibitory receptors belonging to the immunoglobulin-like transcript family (ILT). In this context, we investigated the pathways involved in the control of cell-cycle entry of T
Lei Jiang et al.
Molecular medicine reports, 12(5), 7526-7530 (2015-10-16)
The serine/threonine kinases, Rho‑associated protein kinase I and II (ROCK I and II), regulate the cytoskeleton by acting downstream of the small GTPase, Rho, and have been implicated in tumorigenesis and cancer metastasis. Inhibition of ROCK signaling has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service