Skip to Content
Merck
All Photos(1)

Key Documents

A236

Sigma-Aldrich

AB-MECA

solid

Synonym(s):

N6-(4-Aminobenzyl)-9-[5-(methylcarbonyl)-β-D-ribofuranosyl]adenine, N6-(4-Aminobenzyl)-N-methylcarboxamidoadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N7O4
CAS Number:
Molecular Weight:
399.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

off-white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.3 mg/mL
ethanol: 1 mg/mL
DMSO: 10 mg/mL
dilute aqueous base: 3.5 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NCc4ccc(N)cc4)ncnc23

InChI

1S/C18H21N7O4/c1-20-17(28)14-12(26)13(27)18(29-14)25-8-24-11-15(22-7-23-16(11)25)21-6-9-2-4-10(19)5-3-9/h2-5,7-8,12-14,18,26-27H,6,19H2,1H3,(H,20,28)(H,21,22,23)/t12-,13+,14-,18+/m0/s1

InChI key

LDYMCRRFCMRFKB-MOROJQBDSA-N

Gene Information

Biochem/physiol Actions

High affinity A3 adenosine receptor agonist.

Legal Information

Sold under license from the National Institutes of Health.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Olah et al.
Molecular pharmacology, 45(5), 978-982 (1994-05-01)
The rat A3 adenosine receptor (AR) is a recently characterized AR subtype cloned from testis and brain cDNA libraries. N6-2-(4-Amino-3-[125I]iodophenyl)ethyladenosine, a high affinity A1AR agonist, has served as the only radioligand available for study of the A3AR. The relatively low
K A Jacobson et al.
FEBS letters, 336(1), 57-60 (1993-12-20)
The behavioral effects of a selective A3 adenosine receptor agonist 3-IB-MECA (N6-(3-iodobenzyl)-5'-N-methylcarboxamidoadenosine) in mice and the localization of radioligand binding sites in mouse brain were examined. Low levels of A3 adenosine receptors were detected in various regions of the mouse
C Gallo-Rodriguez et al.
Journal of medicinal chemistry, 37(5), 636-646 (1994-03-04)
Adenosine analogues modified at the 5'-position as uronamides and/or as N6-benzyl derivatives were synthesized. These derivatives were examined for affinity in radioligand binding assays at the newly discovered rat brain A3 adenosine receptor and at rat brain A1 and A2a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service