Skip to Content
Merck

Skip To

A2189

Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase, Mouse monoclonal

clone MB-11, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.2 mL
Check Cart for Availability
$283.00

About This Item

NACRES:
NA.77
UNSPSC Code:
12352203
Conjugate:
alkaline phosphatase conjugate
Clone:
MB-11, monoclonal
Application:
DB, ELISA (d)
Citations:
9

$283.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

alkaline phosphatase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

secondary antibodies

clone

MB-11, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:35,000, dot blot: 1:20,000 using 20 ng of human IgM/dot, dot blot: 1:30,000 using 20 ng of human IgM/dot (chemiluminescence)

isotype

IgG2b

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Human IgMs are the initial immunoglobulin isotypes that appear in blood in response to the first exposure to external antigens. IgMs have been implicated in CNS myelin repair and increased secretory IgM levels have been linked to rhino-conjunctivitis . Monoclonal Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody is specific for human IgM. The product does not react with human IgG, IgA or light chains.
Monoclonal Anti-Human IgM (mouse IgG2b isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Immunogen

Human IgM isolated from pooled normal human serum

Application

Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody purified from hybridoma cell culture has been used in immunocytochemistry and enzyme linked immunosorbent assay (ELISA)

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2064A3076A9688
biological source

mouse

biological source

mouse

biological source

mouse

biological source

goat

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse

technique(s)

direct ELISA: 1:35,000, dot blot: 1:30,000 using 20 ng of human IgM/dot (chemiluminescence), dot blot: 1:20,000 using 20 ng of human IgM/dot

technique(s)

direct ELISA: 1:50,000, dot blot: 1:80,000, immunohistochemistry: suitable, western blot (chemiluminescent): 1:80,000

technique(s)

direct ELISA: 1:6,000, dot blot: 1:1,000

technique(s)

direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000

antibody form

purified from hybridoma cell culture

antibody form

purified immunoglobulin

antibody form

purified from hybridoma cell culture

antibody form

affinity isolated antibody

isotype

IgG2b

isotype

IgG1

isotype

IgG2b

isotype

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters