Skip to Content
Merck
All Photos(1)

Key Documents

A2042

Sigma-Aldrich

Arg-Lys-Glu-Val-Tyr acetate salt

≥97% (HPLC)

Synonym(s):

SP-5, Splenin Fragment 32-36, Splenopentin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H51N9O9
CAS Number:
Molecular Weight:
693.79
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

Gene Information

mouse ... TAC1(281512)

Biochem/physiol Actions

Active fragment of bovine splenin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Audhya et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(9), 2847-2849 (1984-05-01)
Thymopoietin, a 49 amino acid polypeptide hormone of the thymus discovered by its effect on neuromuscular transmission, was later shown to induce T-cell differentiation and to affect immunoregulatory balance. A radioimmunoassay for thymopoietin revealed a crossreaction with a product found
Endika Haro et al.
PLoS genetics, 10(8), e1004468-e1004468 (2014-08-29)
The formation and maintenance of the apical ectodermal ridge (AER) is critical for the outgrowth and patterning of the vertebrate limb. The induction of the AER is a complex process that relies on integrated interactions among the Fgf, Wnt, and
Tatsuya Tsukui et al.
The American journal of pathology, 185(11), 2939-2948 (2015-10-13)
Pulmonary fibrosis is a devastating disease for which there are few effective therapies. Activated fibroblasts form subepithelial clusters known as fibroblastic foci, which are characterized by excessive collagen deposition. The origin of activated fibroblasts is controversial and needs to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service