Skip to Content
Merck
All Photos(2)

Key Documents

A1852

Sigma-Aldrich

Adenosine 5′-triphosphate (ATP) disodium salt hydrate

vial of 30 mg

Synonym(s):

ATP disodium hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$92.70

$92.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$92.70

About This Item

Linear Formula:
C10H14N5O13P3Na2 · xH2O
CAS Number:
Molecular Weight:
551.14 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

$92.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

microbial (Corynebacterium sp.)

Assay

≥99% (HPLC)

form

powder

packaging

vial of 30 mg

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

NTBQNWBHIXNPRU-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine 5γ-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger CAMP. ATP provides the metabolic energy to drive metabolic pumps.

Application

ATP serves as a coenzyme in a wide array of enzymatic reactions.Adenosine 5′-triphosphate (ATP) disodium salt hydrate is used for tissue preparation and maintenance.[1]
Adenosine 5′-triphosphate (ATP) disodium salt hydrate has been used:
  • in rat hippocampal slice cultures to mimic synaptic signalling[2]
  • in the isolation of plasmid from primary porcine alveolar macrophages (PAMs)[3]
  • as a component of hot reaction kinase buffer for in gel kinase assay[4]

Biochem/physiol Actions

P2 purinergic agonist; increases activity of Ca2+-activated K+ channels; substrate for ATP-dependent enzyme systems

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark D A van Logtestijn et al.
PloS one, 10(2), e0117292-e0117292 (2015-02-12)
The stratum corneum (SC) provides a permeability barrier that limits the inflow and outflow of water. The permeability barrier is continuously and dynamically formed, maintained, and degraded along the depth, from the bottom to the top, of the SC. Naturally
Bio-Protocol
Pizzio GA and Rodriguez PL
Testing, 7(5) (2017)
NS1 protein of 2009 pandemic influenza A virus inhibits porcine NLRP3 inflammasome-mediated interleukin-1 beta production by suppressing ASC ubiquitination
Park HS, et al.
Journal of Virology, JVI-00022 (2018)
JuanJuan Duan et al.
Cellular signalling, 58, 44-52 (2019-03-12)
Persistent inflammasome activation contributes to chronic, low grade inflammation. However, it is unclear how the inflammasome activation is sustained after initiation. Here we reported that N4-acetylcytidine (N4A), a nucleoside metabolite, activated microglia and sustained NLRP3 inflammasome activation by inducing HMGB1
Behnam Abbasian et al.
mSphere, 4(5) (2019-09-06)
Urgency urinary incontinence (UUI) and overactive bladder (OAB) can both potentially be influenced by commensal and urinary tract infection-associated bacteria. The sensing of bladder filling involves interplay between various components of the nervous system, eventually resulting in contraction of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service