Skip to Content
Merck
All Photos(1)

Documents

A1151

Sigma-Aldrich

Anti-Albumin antibody produced in goat

fractionated antiserum, lyophilized

Synonym(s):

Albumin Detection Antibody, Anti-Albumin, Goat Anti-Albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

recombinant

expressed in goat

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

mol wt

antigen 65 kDa

species reactivity

human

packaging

vial of 2 mL lyophilized antiserum

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... ALB(213)

Related Categories

General description

Albumin, the major protein produced by liver cells, represents more than half of the total protein content of human serum. Many other body fluids also contain albumin. Three major functions for serum albumin have been proposed: maintenance of osmotic pressure, transportation of a variety of substances and an endogenous source of amino acids. The primary sites of albumin degradation are not known, but the protein can be metabolized by almost every organ in the body. Determination of serum albumin levels is a widely used screening test in clinical medicine. A decrease in serum albumin levels may indicate disease states such as malnutrition, cirrhosis, nephrotic syndrome, diabetes, gastrointestinal and hepatic diseases, thermal burns and pulmonary disease.
Goat polyclonal anti-Albumin antiserum is immunospecific for human albumin by immunoelectrophoresis against normal human serum and human albumin. Identity and purity of the specific antibody is established by immunoelectrophoresis (IEP). Electrophoresis of the antiserum followed by diffusion against anti-goat serum results in multiple arcs of precipitation, whereas against anti-goat IgG, it results in a single arc of precipitation in the γ region.

Immunogen

purified human albumin

Application

Goat polyclonal anti-Albumin antiserum is used to tag albumin for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of albumin a variety of clinical indications.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elliott L Rodriguez et al.
Journal of chromatography. A, 1638, 461683-461683 (2020-11-24)
Diabetes is characterized by elevated levels of blood glucose, which can result in the modification of serum proteins. The modification of a protein by glucose, or glycation, can also lead to the formation of advanced glycated end-products (AGEs). One protein
Ryan Matsuda et al.
Journal of chromatography. A, 1416, 112-120 (2015-09-19)
A method combining on-line immunoextraction microcolumns with high-performance affinity chromatography (HPAC) was developed and tested for use in examining drug-protein interactions with normal or modified proteins. Normal human serum albumin (HSA) and glycated HSA were used as model proteins for
Sadia Sharmeen et al.
Current protocols, 3(8), e867-e867 (2023-08-23)
Immunoaffinity chromatography (IAC) is a type of liquid chromatography that uses immobilized antibodies or related binding agents as selective stationary phases for sample separation or analysis. The strong binding and high selectivity of antibodies have made IAC a popular tool
Elliott L Rodriguez et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1136, 121812-121812 (2019-12-17)
An on-line purification and entrapment system was developed that could extract a protein from a sample such as serum and entrap this protein within a small column for use in high-performance affinity chromatography. Human serum albumin (HSA) was employed as
G Grossmann et al.
Journal of applied physiology (Bethesda, Md. : 1985), 82(6), 2003-2010 (1997-06-01)
Near-term newborn rabbits were exposed via the airways to a monoclonal antibody to surfactant protein B and ventilated for 0-120 min. Control animals received nonspecific rabbit or mouse immunoglobulin G, saline, or no material via the airways. Administration of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service