Skip to Content
Merck
All Photos(1)

Key Documents

A1049

Sigma-Aldrich

(±)-Abscisic acid

suitable for plant cell culture, BioReagent, ≥98.5% (HPLC)

Synonym(s):

(2Z,4E)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-2,4-pentadienoic acid, ABA, Dormin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

≥98.5% (HPLC)

form

powder

technique(s)

cell culture | plant: suitable

mp

186-188 °C (lit.)

solubility

methanol: 50 mg/mL, clear to slightly hazy

application(s)

agriculture

storage temp.

−20°C

SMILES string

CC(\C=C\C1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-

InChI key

JLIDBLDQVAYHNE-LXGGSRJLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Abscisic acid (ABA) is an isoprenoid plant hormone (phytohormone) synthesized in all parts of plants and involved in many plant developmental processes. ABA is involved in establishing dormancy and regulation of stress responses. ABA inhibits fruit ripening, seed germination, photosynthesis and kinetin biosynthesis.

Preparation Note

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hormone Crosstalk in Plant Disease and Defense: More Than Just JASMONATE-SALICYLATE Antagonism.
Robert-Seilaniantz A, Grant M, Jones JD.
Annual review of phytopathology (2010)
Zuying Zhang et al.
Plant biotechnology journal, 18(11), 2267-2279 (2020-03-28)
The RAV (related to ABI3/viviparous 1) group of transcription factors (TFs) play multifaceted roles in plant development and stress responses. Here, we show that strawberry (Fragaria × ananassa) FaRAV1 positively regulates anthocyanin accumulation during fruit ripening via a hierarchy of activation processes.
Xue Yang et al.
Journal of experimental botany, 71(6), 2142-2156 (2019-12-25)
The chloroplast protein ferredoxin 1 (FD1), with roles in the chloroplast electron transport chain, is known to interact with the coat proteins (CPs) of Tomato mosaic virus and Cucumber mosaic virus. However, our understanding of the roles of FD1 in
Role of DREBs in regulation of abiotic stress responses in plants.
Lata C, Prasad M.
Journal of Experimental Botany (2011)
Fabio Pasin et al.
Plant communications, 1(5) (2020-09-29)
A complex network of cellular receptors, RNA targeting pathways, and small-molecule signaling provides robust plant immunity and tolerance to viruses. To maximize their fitness, viruses must evolve control mechanisms to balance host immune evasion and plant-damaging effects. The genus Potyvirus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service