Skip to Content
Merck
All Photos(1)

Key Documents

A0987

Sigma-Aldrich

n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate

≥98% (HPLC)

Synonym(s):

Cytosporone A n-amyl ester, compound 10i

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$581.00

$143.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$143.00
25 MG
$581.00

About This Item

Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$143.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >65 mg/mL

storage temp.

2-8°C

SMILES string

CCCCCCCCC(=O)c1c(O)cc(O)cc1CC(=O)OCCCCC

InChI

1S/C22H34O5/c1-3-5-7-8-9-10-12-19(24)22-17(14-18(23)16-20(22)25)15-21(26)27-13-11-6-4-2/h14,16,23,25H,3-13,15H2,1-2H3

InChI key

FCKDHZVFIMRCNM-UHFFFAOYSA-N

Biochem/physiol Actions

n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate is a Nur77 agonist.
n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate is a Nur77 agonist. Nur77 is a steroid orphan receptor that plays a critical role in regulating proliferation, differentiation and apoptosis. n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate, a cytosporone B analog, is a stronger activator of Nur77 than it parent compound. n-Amyl 2-[3,5-dihydroxy-2-(1-nonanoyl)phenyl]acetate treatment causes Nur77 migration to mitochondria, which results in mitochondrial Cyt c release into the cytoplasm and mitochondrial AIF redistribution in both the cytoplasm and nucleus.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service