A0976
Monoclonal Anti-Angiostatin antibody produced in mouse
clone 79735, purified immunoglobulin, lyophilized powder
Synonym(s):
Anti-Angiostatin Kringle 1-3
About This Item
biological source
mouse
recombinant
expressed in E. coli
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
79735, monoclonal
form
lyophilized powder
mol wt
antigen 38 kDa
species reactivity
human
should not react with
mouse
technique(s)
western blot: 1-2 μg/mL
isotype
IgG2a
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PLG(5340)
General description
Monoclonal Anti-Angiostatin recognizes human angiostatin. This antibody also recognizes plasminogen but shows no cross-reactivity with recombinant mouse angiostatin.
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service