A0877
L-Aspartic acid di-tert-butyl ester hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H23NO4 · HCl
CAS Number:
Molecular Weight:
281.78
Beilstein:
5652807
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Assay
≥98%
form
powder
color
white
storage temp.
−20°C
SMILES string
Cl.CC(C)(C)OC(=O)C[C@H](N)C(=O)OC(C)(C)C
InChI
1S/C12H23NO4.ClH/c1-11(2,3)16-9(14)7-8(13)10(15)17-12(4,5)6;/h8H,7,13H2,1-6H3;1H/t8-;/m0./s1
InChI key
GVLZIMQSYQDAHB-QRPNPIFTSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester (G7501).
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
W J Freed et al.
Neurobehavioral toxicology and teratology, 7(3), 275-278 (1985-05-01)
Glutamic acid di-tert butyl ester (GTBE) was found to have a pronounced convulsant effect in mice and rats, producing recurrent clonic convulsions combined with postural and respiratory disturbances in a dosage of 0.5 mmol/kg (148 mg/kg). Tert-butyl ester derivatives of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service