Skip to Content
Merck
All Photos(1)

Key Documents

A0796

Sigma-Aldrich

p-Aminomethylbenzenesulfonamide–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

form

saline suspension

extent of labeling

≥6 mg per mL

matrix

crosslinked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

p-Aminomethylbenzenesulfonamide is an agarose in saline suspension that can be used in affinity chromatography, protein chromatography and specialty resins. p-Aminomethylbenzenesulfonamide has been used in studies assessing inhibition of mitochondrial carbonic anhydrase and ureagenesis.

Physical form

Suspension in 0.5 M NaCl containing preservative.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Dodgson et al.
Journal of applied physiology (Bethesda, Md. : 1985), 60(2), 646-652 (1986-02-01)
The amount of urea produced in 60 min, [urea]t = 60, from intact guinea pig hepatocytes incubated in NH4Cl, oleate, lactate, NaHCO3, and ornithine at 37 degrees C at pH 7.1 is decreased by ethoxzolamide (EZ): Ki,EZ [urea]t = 60
B Ulmasov et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(26), 14212-14217 (2000-12-20)
Carbonic anhydrase XII (CA XII) is a transmembrane glycoprotein with an active extracellular CA domain that is overexpressed on cell surfaces of certain cancers. Its expression has been linked to tumor invasiveness. To characterize its catalytic properties, we purified recombinant
D K Srivastava et al.
Journal of the American Chemical Society, 129(17), 5528-5537 (2007-04-05)
Despite the similarity in the active site pockets of carbonic anhydrase (CA) isozymes I and II, the binding affinities of benzenesulfonamide inhibitors are invariably higher with CA II as compared to CA I. To explore the structural basis of this
S Sender et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 47(1), 43-50 (1998-12-19)
Flounders Platichthys flesus were investigated with respect to isolation, purification, and cellular localization of carbonic anhydrase (CA) in the respiratory system. CA was purified from gills and erythrocytes and was shown to exclusively represent a soluble enzyme with an apparent
S J Dodgson
Journal of applied physiology (Bethesda, Md. : 1985), 63(5), 2134-2141 (1987-11-01)
The amount of urea synthesized in intact guinea pig hepatocytes in 60 min ([urea]t=60), was determined at 37 degrees C in Krebs-Henseleit buffer plus (in mM) 10 NH4Cl, 5 lactate, and 10 ornithine in 5% CO2-95% O2. The concentrations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service