A024
MECA
solid
Synonym(s):
5′-N-Methylcarboxamidoadenosine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H14N6O4
CAS Number:
Molecular Weight:
294.27
MDL number:
UNSPSC Code:
12352200
Recommended Products
form
solid
color
white
solubility
0.1 M HCl: soluble
H2O: slightly soluble
ethanol: slightly soluble
SMILES string
CNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23
Gene Information
human ... ADORA2A(135) , ADORA2B(136)
Biochem/physiol Actions
A2 adenosine receptor agonist.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R F Bruns et al.
Molecular pharmacology, 29(4), 331-346 (1986-04-01)
[3H]NECA (1-(6-amino-9H-purin-9-yl)-1-deoxy-N-ethyl-beta-D-ribofuronamide) is known to bind to both the A1 and A2 subtypes of adenosine receptor in rat striatal membranes. In order to study the putative A2 component of [3H]NECA binding, we examined several compounds for the ability to selectively
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service