Skip to Content
Merck
All Photos(1)

Documents

Y1376

Sigma-Aldrich

Yeast Synthetic Drop-out Medium Supplements

without leucine

Synonym(s):

Synthetic Yeast Growth Media

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
ND.02

Quality Level

form

powder

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The selection of plasmids in yeast is based on the use of auxotrophic mutant strains that cannot grow without a specific media component (an amino acid, purine or pyrimidine). Transformation with a plasmid containing the mutated gene enables the transformant to grow on a medium lacking the required component. Sigma′s Yeast Synthetic Drop-Out Media Supplements create a richer medium for better yield and growth rate, and increase the probability of successful transformations when screening libraries or performing gene knock-outs.

Components

The supplements contain all of the following components except as indicated:
Amino acids: All standard amino acids except for leucine. All the amino acids are present at a concentration of 76 mg/L.
Other nutrients: Adenine (18 mg/L), inositol (76 mg/L), p-aminobenzoic acid (8 mg/L), uracil (76 mg/L).

Other Notes

Mixtures of amino acids and other nutrients to be added to Yeast Nitrogen Base Without Amino Acids.

Quantity

1.6 g will supplement 1 liter of medium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Snoek et al.
Nucleic acids research, 48(1), e3-e3 (2019-11-30)
Allosteric transcription factors (aTFs) have proven widely applicable for biotechnology and synthetic biology as ligand-specific biosensors enabling real-time monitoring, selection and regulation of cellular metabolism. However, both the biosensor specificity and the correlation between ligand concentration and biosensor output signal
Renata Novakova et al.
Applied microbiology and biotechnology, 102(2), 857-869 (2017-12-03)
Mithramycin A is an antitumor compound used for treatment of several types of cancer including chronic and acute myeloid leukemia, testicular carcinoma, hypercalcemia and Paget's disease. Selective modifications of this molecule by combinatorial biosynthesis and biocatalysis opened the possibility to
Christopher J Wilkinson et al.
BMC cell biology, 10, 17-17 (2009-03-04)
The centrosome is the cell's microtubule organising centre, an organelle with important roles in cell division, migration and polarity. However, cells can divide and flies can, for a large part of development, develop without them. Many centrosome proteins have been

Articles

Genetic engineering enables large-scale expression and isolation of recombinant proteins for research purposes.

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service