Skip to Content
Merck
All Photos(1)

Key Documents

C8302

Sigma-Aldrich

4-Chloro-1-naphthol solution

For HRP detection on Western Blots

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$168.00

$168.00


Estimated to ship onMarch 27, 2025



Select a Size

Change View
100 ML
$168.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.52

$168.00


Estimated to ship onMarch 27, 2025


grade

for molecular biology

Quality Level

form

liquid

storage temp.

2-8°C

SMILES string

Oc1ccc(Cl)c2ccccc12

InChI

1S/C10H7ClO/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,12H

InChI key

LVSPDZAGCBEQAV-UHFFFAOYSA-N

General description

4-Chloro-1-naphthol is a substrate for HRP in a reaction that results in a colored precipitate. It is commonly used for colorimetric detection and visualization of proteins in western blotting.

Application

Suitable for visualizing protein-peroxidase conjugates in western blotting.

Components

This product contains 0.48 mM 4-chloro-1-naphthol, 50 mM Tris-HCl, 0.2 M NaCl and 17% methanol.

Other Notes

0.48 mM 4-chloro-1-naphthol, 50 mM Tris-HCl, 0.2 M NaCl and 17% methanol.

Principle

Enzymes such as HRP convert 4-Chloro-1-naphthol to a purple-colored precipitate on the blot that is readily visible to the eye.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

163.9 °F - closed cup

Flash Point(C)

73.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hwan-Su Hwang et al.
Planta, 252(3), 44-44 (2020-09-03)
Overexpression of the tobacco lipid transfer protein (NtLTP1) gene in transgenic orange mint resulted in enhanced accumulation of monoterpenes in the cavity of head cells of glandular trichomes, which resulted in enhanced emission of monoterpenes from transgenic orange mints. Plants
Animesh Chowdhury et al.
Cell biology international, 44(5), 1142-1155 (2020-01-23)
We sought to determine the mechanism by which angiotensin II (AngII) inhibits isoproterenol induced increase in adenylate cyclase (AC) activity and cyclic adenosine monophosphate (cAMP) production in bovine pulmonary artery smooth muscle cells (BPASMCs). Treatment with AngII stimulates protein kinase

Articles

This solution is used as a substrate for detecting horseradish-peroxidase conjugates in Western blotting.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service