Skip to Content
Merck
All Photos(1)

Documents

94124

Sigma-Aldrich

sn-Glycerol 3-phosphate lithium salt

≥95.0% (TLC)

Synonym(s):

D-Glycerol 1-phosphate lithium salt, L-Glycerol 3-phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9O6P · xLi+
CAS Number:
Molecular Weight:
172.07 (free acid basis)
UNSPSC Code:
12352211
NACRES:
NA.25

Quality Level

Assay

≥95.0% (TLC)

form

solid

optical activity

[α]/D +2.0±0.5°°, c = 1 in H2O

lipid type

phosphoglycerides

storage temp.

−20°C

InChI

1S/C3H9O6P/c4-1-3(5)2-9-10(6,7)8/h3-5H,1-2H2,(H2,6,7,8)/t3-/m1/s1

InChI key

AWUCVROLDVIAJX-GSVOUGTGSA-N

Biochem/physiol Actions

Glycerol 3-phosphate (G3P) is an important intermediate of carbohydrate and lipid metabolic pathways. It is produced from glycerol by glycerol kinase or from dihydroxyacetone phosphate by glycerol 3-phosphate dehydrogenase. G3P may enter the G3P shuttle to generate NAD+, or may be converted to glyceraldehyde 3-phosphate and enter glycolysis or the lipid biosynthesis pathway. Ugp-dependent transport system for G3P in E. coli.
Metabolite in glycerolipid metabolism and glycerophospholipid metabolism. Ugp-dependent transport system for G3P in E. coli.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L André et al.
FEBS letters, 286(1-2), 13-17 (1991-07-29)
Production of glycerol and a key enzyme in glycerol production, glycerol 3-phosphate dehydrogenase (NAD+) (GPD), was studied in Saccharomyces cerevisiae cultured in basal media or media of high salinity, with glucose, raffinose or ethanol as the sole carbon source. At
J Norbeck et al.
The Journal of biological chemistry, 271(23), 13875-13881 (1996-06-07)
The existence of specific dl-glycerol-3-phosphatase (EC 3.1.3.21) activity in extracts of Saccharomyces cerevisiae was confirmed by examining strains lacking nonspecific acid and alkaline phosphatase activities. During purification of the glycerol-3-phosphatase, two isozymes having very similar molecular weights were isolated by
H Schweizer et al.
Journal of bacteriology, 150(3), 1154-1163 (1982-06-01)
The ugp-dependent transport system for sn-glycerol-3-phosphate has been characterized. The system is induced under conditions of phosphate starvation and in mutants that are constitutive for the pho regulon. The system does not operate in membrane vesicles and is highly sensitive
Won-Ki Huh et al.
Nature, 425(6959), 686-691 (2003-10-17)
A fundamental goal of cell biology is to define the functions of proteins in the context of compartments that organize them in the cellular environment. Here we describe the construction and analysis of a collection of yeast strains expressing full-length
Concentrations of intermediary metabolites in yeast.
J M Gancedo et al.
Biochimie, 55(2), 205-211 (1973-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service