Skip to Content
Merck
All Photos(1)

Key Documents

93658

Sigma-Aldrich

Esterase Pyrobaculum calidifontis, recombinant

recombinant, expressed in E. coli, ≥2.0 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

crystalline
crystals
powder or flakes

specific activity

≥2.0 U/mg

storage temp.

−20°C

Application

Esterase, from Pyrobaculum calidifontis, may be used in structural biology research[1].

Biochem/physiol Actions

An esterase is a hydrolase that splits esters into acids and alcohols.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gottfried J Palm et al.
Applied microbiology and biotechnology, 91(4), 1061-1072 (2011-05-27)
The highly thermostable esterase from the hyperthermophilic archaeon Pyrobaculum calidifontis VA1 (PestE) shows high enantioselectivity (E > 100) in the kinetic resolution of racemic chiral carboxylic acids, but little selectivity towards acetates of tertiary alcohols (E = 2-4). To explain
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine
Joshua P Lewis et al.
Pharmacogenetics and genomics, 23(1), 1-8 (2012-11-01)
Carboxylesterase 1 (CES1) is the primary enzyme responsible for converting clopidogrel into biologically inactive carboxylic acid metabolites. We genotyped a functional variant in CES1, G143E, in participants of the Pharmacogenomics of Anti-Platelet Intervention (PAPI) study (n=566) and in 350 patients
Judit Marsillach et al.
Toxicology, 307, 46-54 (2012-12-25)
Exposure to organophosphorus (OP) compounds can lead to serious neurological damage or death. Following bioactivation by the liver cytochromes P450, the OP metabolites produced are potent inhibitors of serine active-site enzymes including esterases, proteases and lipases. OPs may form adducts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service