Skip to Content
Merck
All Photos(1)

Documents

93328

Sigma-Aldrich

Trizma® maleate

BioUltra, ≥99.5% (NT)

Synonym(s):

TRIS maleate salt, Tris(hydroxymethyl)aminomethane maleate salt, Tromethamine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · C4H4O4
CAS Number:
Molecular Weight:
237.21
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.5% (NT)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue (900 °C)

≤0.1% (as SO4)

pH

3.0-4.5 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 328 nm in H2O at 0.5 M

SMILES string

NC(CO)(CO)CO.OC(=O)\C=C/C(O)=O

InChI

1S/C4H11NO3.C4H4O4/c5-4(1-6,2-7)3-8;5-3(6)1-2-4(7)8/h6-8H,1-3,5H2;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

HTMWOUBCEZXSHN-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trizma is used in the formulation of buffer solutions in the pH range between 7.5 and 8.5. Tris buffer solutions are widely used in cell and molecular biology for processes such as protein and nucleic acid extraction and purification. Trizma based buffers are also in column chromatography and in gel electrophoresis. Trizma maleate is used in the preparation of Tris maleate buffers with lower effective pH buffering capacity than Tris buffers. Tris maleate buffers may be used in various grow media, in tissue preparations and as a diluent or washing buffer.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Chikayama et al.
Histochemistry and cell biology, 113(6), 489-495 (2000-08-10)
We investigated the enzyme cytochemical localization of sarcosine oxidase (SOX) in the liver and kidney of several mammals using a cerium technique. First we measured the enzyme activities in the liver and kidney of several mammals and in several organs
Tamás Juhász et al.
Applied biochemistry and biotechnology, 121-124, 243-254 (2005-05-27)
The hydrolysis of cellulose to the water-soluble products cellobiose and glucose is achieved via synergistic action of cellulolytic proteins. The three types of enzymes involved in this process are endoglucanases, cellobiohydrolases, and beta-glucosidases. One of the best fungal cellulase producers
A D Yaremchuk et al.
Proteins, 21(3), 261-264 (1995-03-01)
Lysyl-tRNA synthetase from Thermus thermophilus has been cocrystallized with either its cognate tRNAlys or Escherichia coli tRNAlys using ammonium sulfate as precipitant. The crystals grow from solutions containing a 1:2.5 stoichiometry of synthetase dimer to tRNA in 18-22% ammonium sulfate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service