Peroxide Block is a 3% hydrogen peroxide solution commonly used for performing IHC.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
description
[For In Vitro Diagnostic Use in Select Regions]
form
aqueous solution
packaging
pkg of 1 L (925B-09), pkg of 100 mL ready to use (925B-04), pkg of 15 mL ready to use (925B-02), pkg of 200 mL ready to use (925B-05), pkg of 50 mL ready to use (925B-03)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry: suitable
shipped in
ambient
General description
Physical form
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| shipped in ambient | shipped in - | shipped in - | shipped in wet ice |
| form aqueous solution | form - | form lyophilized powder | form lyophilized |
| packaging pkg of 1 L (925B-09), pkg of 15 mL ready to use (925B-02), pkg of 50 mL ready to use (925B-03), pkg of 100 mL ready to use (925B-04), pkg of 200 mL ready to use (925B-05) | packaging kit of 1 (7 components) | packaging - | packaging pkg of 8 mg (~40 mg lyophilizate) |
| technique(s) immunohistochemistry: suitable | technique(s) ELISA: suitable, immunoblotting: suitable, immunohistochemistry: suitable | technique(s) direct ELISA: 1:80,000 using human IgG | technique(s) - |
| Quality Level 500 | Quality Level - | Quality Level 100, 200 | Quality Level - |
| description [For In Vitro Diagnostic Use in Select Regions] | description - | description - | description - |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
There is a requirement for large volumes of H2O2 (containers that exceed 1000 mls), please suggest
1 answer-
Helpful?
-
-
What is the % of peroxide in peroxide block 925B-09?
1 answer-
a 3% peroxide block solution should be used.
Helpful?
-
-
Can peroxide block as a diluent or how is it used?
1 answer-
The peroxide block is used to neutralize the endogenous peroxidase that arises from using an HRP or Horse Radish Peroxidase kit. It is part of the protocol and is typically used for 10 minutes as one of the steps.
Helpful?
-


