Skip to Content
Merck
All Photos(1)

Key Documents

92521

Sigma-Aldrich

L-Threonic acid lithium salt

≥98.5% (TLC)

Synonym(s):

(2R,3S)-2,3,4-Trihydroxybutyric acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O5 · xLi+
CAS Number:
Molecular Weight:
136.10 (free acid basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

≥98.5% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

SMILES string

OC[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C4H8O5/c5-1-2(6)3(7)4(8)9/h2-3,5-7H,1H2,(H,8,9)/t2-,3+/m0/s1

InChI key

JPIJQSOTBSSVTP-STHAYSLISA-N

Biochem/physiol Actions

Metabolite of ascorbate and aldarate metabolism, substrate of L-threonate 3- dehydrogenase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Li et al.
Molecular brain, 7, 65-65 (2014-09-13)
Profound synapse loss is one of the major pathological hallmarks associated with Alzheimer's disease, which might underlie memory impairment. Our previous work demonstrates that magnesium ion is a critical factor in controlling synapse density/plasticity. Here, we tested whether elevation of
Absolute Configuration of β- and α-Asymmetric Carbons within β-O-4-Structures in Hardwood Lignin.
Akiyama, T., et al.
J. Wood Chem. Technol., 35, 8-16 (2014)
S N Reinke et al.
Multiple sclerosis (Houndmills, Basingstoke, England), 20(10), 1396-1400 (2014-01-29)
Metabolomics enables the provision of sensitive bio-markers of disease. We performed 800 MHz (1)H-nuclear magnetic resonance (NMR) spectroscopic analyses of cerebrospinal fluid (CSF) specimens to identify biomarkers of multiple sclerosis (MS), yielding reproducible detection of 15 metabolites from MS (n=15)
Lianzhen Yu et al.
Journal of gastroenterology and hepatology, 26(8), 1290-1297 (2011-03-30)
To study the low-molecular-weight metabolites in blood plasma of patients with the progressive disease, gastric cancer, and to characterize different stages from chronic superficial gastritis (CSG) to chronic atrophic gastritis (CAG), intestinal metaplasia (IM), gastric dysplasia (DYS) and finally gastric
R W Watts et al.
Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Beiheft, 15, 216-225 (1976-01-01)
The excretion of urinary acidic metabolites by 10 patients undergoing gastric or biliary tract surgery has been studied. Five patients were infused with xylitol and 5 with glucose. Four of the xylitol-infused patients had hyperglycollic aciduria and 3 of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service