Skip to Content
Merck
All Photos(1)

Key Documents

91893

Sigma-Aldrich

Poly(ethylene glycol)

BioUltra, 400

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$69.10
1 L
$153.00

$69.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$69.10
1 L
$153.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26

$69.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

BioUltra

form

liquid

mol wt

Mr 380-420

impurities

insoluble matter:, passes filter test
≤0.02% peroxides (as H2O2):

refractive index

n20/D 1.467

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL at 25 °C, clear, colorless

density

1.126 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤300 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.03

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a non-ionic hydrophilic polymer and is available in different molecular weights. It facilitates the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. PEG also interacts with cell membrane, thereby allowing cell fusion.[1]

Application

Poly(ethylene glycol) (PEG) has been used to dissolve JC-124, nucleotide-binding domain, leucine rich family pyrin containing 3 gene (NLRP3) inflammasome inhibitor[2] and solubilize CoQ10, a mitochondrial coenzyme Q4.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenhai Sui et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(22), 10937-10942 (2019-05-16)
Mirabegron (Myrbetriq) is a β3-adrenoreceptor agonist approved for treating overactive bladder syndrome in human patients. This drug can activate brown adipose tissue (BAT) in adult humans and rodents through the β3-adrenoreceptor-mediated sympathetic activation. However, the effect of the mirabegron, approved
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
NLRP3 inflammasome inhibitor ameliorates amyloid pathology in a mouse model of Alzheimer?s disease
Yin J, et al.
Molecular Neurobiology, 55(3), 1977-1987 (2018)
Woori Jo et al.
International journal of molecular sciences, 21(18) (2020-09-25)
The incidence of myocardial infarction, among the causes of cardiovascular morbidity and mortality, is increasing globally. In this study, left ventricular (LV) dysfunction, including LV systolic and diastolic function, was investigated in a rat myocardial ischemia/reperfusion injury model with echocardiography.
Idebenone and coenzyme Q10 are novel PPAR alpha gamma ligands, with potential for treatment of fatty liver diseases
Tiefenbach J, et al.
Disease models & mechanisms, 11(9), dmm034801-dmm034801 (2018)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service