Skip to Content
Merck
All Photos(1)

Key Documents

90699

Sigma-Aldrich

Fluorescent IEF-Marker pI 9.0 solution

1 mg/mL in 10 mM HCl

Synonym(s):

IEF-Marker fluorescent pI 9.0

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$361.00

$361.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$361.00

About This Item

MDL number:
UNSPSC Code:
41105336
NACRES:
NA.26

$361.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

mol wt

Mw 370.49 g/mol

concentration

1 mg/mL in 10 mM HCl

fluorescence

λex 385 nm; λem 495 nm in 0.1 M Tris pH 9.0

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

2-8°C

General description

filtered through a 0.45 μm membrane filter

Application

suitable as IEF-Marker for HPCE and for IEF-gels

Reconstitution

For use in HPCE the marker should be diluted 1:100 in a suitable ampholyte solution (e.g. 2% ampholyte 3-10), UV-detection at 280 nm (20 °C). For use in IEF-gels the solution can be loaded directly onto the gel

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service