Skip to Content
Merck
All Photos(1)

Key Documents

90572

Sigma-Aldrich

D-Galactose 4-sulfate sodium salt

≥97.0% (TLC)

Synonym(s):

Gal-4S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O9SNa
CAS Number:
Molecular Weight:
282.20
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97.0% (TLC)

storage temp.

−20°C

SMILES string

[Na+].OC[C@@H](O)[C@H](OS([O-])(=O)=O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O9S.Na/c7-1-3(9)5(11)6(4(10)2-8)15-16(12,13)14;/h1,3-6,8-11H,2H2,(H,12,13,14);/q;+1/p-1/t3-,4+,5+,6-;/m0./s1

InChI key

PPFRJSVPFMKACS-NQZVPSPJSA-M

Looking for similar products? Visit Product Comparison Guide

Application

D-Galactose 4-sulfate (Gal-4S) may be used to help differentiate glycan sulfatide recognition/binding sites and to study the structure and biochemisty of carrageenans.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Yamada et al.
European journal of biochemistry, 233(2), 687-693 (1995-10-15)
The carbohydrate-protein linkage region of a chondroitin 4-sulfate chain attached to urinary trypsin inhibitor (UTI) was isolated from human urine and characterized structurally. The chondroitin 4-sulfate chain was released from UTI by beta-elimination using alkaline NaBH4 then digested with chondroitinase
Rando Tuvikene et al.
Carbohydrate research, 344(6), 788-794 (2009-03-10)
The composition, structure, and thermal stability of carrageenans from unattached Coccotylus truncatus (the Baltic Sea, Estonia) were investigated. The complex polysaccharide was characterized by (13)C NMR and FTIR spectroscopy, ICP-OES and gel permeation chromatography methods. The main components of C.
T Barbeyron et al.
The Journal of biological chemistry, 275(45), 35499-35505 (2000-08-10)
iota-Carrageenases are polysaccharide hydrolases that cleave the beta-1,4 linkages between the d-galactose-4-sulfate and 3, 6-anhydro-d-galactose-2-sulfate residues in the red algal galactans known as iota-carrageenans. We report here on the purification of iota-carrageenase activity from the marine bacterium Zobellia galactanovorans and
Lena Jansson et al.
PloS one, 4(2), e4487-e4487 (2009-02-27)
The first step in the pathogenesis of enterotoxigenic Escherichia coli (ETEC) infections is adhesion of the bacterium to the small intestinal epithelium. Adhesion of ETEC is mediated by a number of antigenically distinct colonization factors, and among these, one of
Astrid Von Mentzer et al.
Virulence, 11(1), 381-390 (2020-04-05)
The ability to adhere via colonization factors to specific receptors located on the intestinal mucosa is a key virulence factor in enterotoxigenic Escherichia coli (ETEC) pathogenesis. Here, the potential glycosphingolipid receptors of the novel human ETEC colonization factor CS30 were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service