Skip to Content
Merck
All Photos(1)

Key Documents

89517

Sigma-Aldrich

(S)-3,4-Dihydroxybutyric acid lithium salt hydrate

≥95.0% (GC)

Synonym(s):

(S)-3,4-Dihydroxybutanoic acid lithium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O4 · xLi+ · yH2O
CAS Number:
Molecular Weight:
120.10 (free acid basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥85% (free acid, qNMR)
≥95.0% (GC)

form

solid

optical activity

[α]/D -25±5°, 24 hr, c = 0.1 in 0.1 M HCl

storage temp.

−20°C

SMILES string

O[C@H](CO)CC(O)=O

InChI

1S/C4H8O4/c5-2-3(6)1-4(7)8/h3,5-6H,1-2H2,(H,7,8)/t3-/m0/s1

InChI key

DZAIOXUZHHTJKN-VKHMYHEASA-N

Biochem/physiol Actions

3,4-Dihydroxybutyric acid is a normal human urinary metabolite that is excreted in increased concentration in patients with succinic semialdehyde dehydro- genase (SSADH) deficiency, while healthy adults excrete 0.37 +/- 0.15 (SD) mmoles of 3,4- dihydroxybutyrate per 24 hr. The renal clearance of this compound is roughly equal that of creatinine. This metabolite is also detectable in blood.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moto-o Nakajima et al.
Rinsho byori. The Japanese journal of clinical pathology, 50(5), 513-518 (2002-06-25)
Urinary free ATP assay by the firefly luciferin-luciferase method is a rapid and simple method for determining renal function, especially uriniferous tubule function. Normal range of urinary free ATP concentration, daily ATP excretion in urine, urinary ATP/creatinine value and ATP
The occurrence of (S)-3,4-dihydroxybutyrate in human blood and urine.
V Fell et al.
Biochemical medicine, 13(1), 40-45 (1975-05-01)
Antonio Noto et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 27 Suppl 2, 46-52 (2014-10-07)
A supervised multivariate model to classify the metabolome alterations between autistic spectrum disorders (ASD) patients and controls, siblings of autistic patients, has been realized and used to realize a network model of the ASD patients' metabolome. In our experiment we
Toshihiro Shinka et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 776(1), 57-63 (2002-07-20)
We describe the rapid and sensitive detection of 4-hydroxybutyric acid, which is a marker compound for succinic semialdehyde dehydrogenase (SSADH) deficiency. Urinary 4-hydroxybutyric acid and 3,4-dihydroxybutyric acid were targeted, quantified by gas chromatography-mass spectrometry after simplified urease digestion in which
Chie Ohdoi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 792(1), 123-130 (2003-06-28)
Lesch-Nyhan syndrome (LNS) is caused by a severe deficiency of hypoxanthine-guanine phosphoribosyltransferase (HPRT) and clinically characterized by self-injurious behavior and nephrolithiasis; the latter is treatable with allopurinol, an inhibitor of xanthine oxidase which converts xanthine and hypoxanthine into uric acid.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service