Skip to Content
Merck
All Photos(1)

Key Documents

89426

Sigma-Aldrich

3-Sulfo-N-succinimidyl benzoate sodium salt

≥99% (HPLC)

Synonym(s):

1-(Benzoyloxy)-2,5-dioxo-3-pyrrolidinesulfonic acid sodium salt, Benzoic acid (3-sulfo-N-succinimidyl) ester sodium salt, Benzoic acid N-hydroxysulfosuccinimide ester sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$195.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 MG
$195.00

About This Item

Empirical Formula (Hill Notation):
C11H8NNaO7S
CAS Number:
Molecular Weight:
321.24
Beilstein:
10311265
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.25

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥99% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)c2ccccc2)C1=O

InChI

1S/C11H9NO7S.Na/c13-9-6-8(20(16,17)18)10(14)12(9)19-11(15)7-4-2-1-3-5-7;/h1-5,8H,6H2,(H,16,17,18);/q;+1/p-1

InChI key

MDKFDOUBUQSEDS-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of Coenzyme A Esters in the Metabolism of Benzoate and Cyclohexanecarboxylate by Rhodopseudomonas palustris.
Georgina N. Hutber
Microbiology, 129, 2413-2420 (1983)
Alexandre Panchaud et al.
Molecular & cellular proteomics : MCP, 7(4), 800-812 (2007-12-18)
Identification and relative quantification of hundreds to thousands of proteins within complex biological samples have become realistic with the emergence of stable isotope labeling in combination with high throughput mass spectrometry. However, all current chemical approaches target a single amino
J V Staros
Biochemistry, 21(17), 3950-3955 (1982-08-17)
We have synthesized and characterized N-hydroxysulfosuccinimide, a new hydrophilic ligand for the preparation of active esters. We have incorporated this ligand into two new protein cross-linking reagents, 3,3'-dithiobis(sulfosuccinimidyl propionate) and bis(sulfosuccinimidyl) suberate. In experiments with rabbit muscle aldolase, it is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service