Skip to Content
Merck
All Photos(1)

Key Documents

89216

Sigma-Aldrich

Pectinase from Aspergillus

≥0.3 U/mg

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$52.80
50 G
$117.00

$52.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$52.80
50 G
$117.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.77

$52.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

Aspergillus sp.

Quality Level

form

powder

specific activity

≥0.3 U/mg

storage temp.

2-8°C

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol galacturonic acid from polygalacturonic acid per minute at pH 4.0 and 50° C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service