Skip to Content
Merck
All Photos(1)

Documents

89126

Sigma-Aldrich

Cetirizin dihydrochloride

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (HPLC)

form

powder

SMILES string

Cl[H].Cl[H].OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

InChI

1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H

InChI key

PGLIUCLTXOYQMV-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Application

Cetirizin (CTZ) may be used in studies on the treatment of cutaneous mastocytosis (urticaria pigmentosa). Cetirizin, an antihistamine, may be useD in allergy research as an antagonist of the histamine 1 receptor (H1R).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gschwandtner et al.
Allergy, 68(1), 37-47 (2012-11-20)
Defects in keratinocyte differentiation and skin barrier are important features of inflammatory skin diseases like atopic dermatitis. Mast cells and their main mediator histamine are abundant in inflamed skin and thus may contribute to disease pathogenesis. Human primary keratinocytes were
K Rossbach et al.
Neuroscience, 190, 89-102 (2011-06-22)
Histamine has long been recognised as a classical inducer of pruritus. However, the specific mechanism of histamine-induced itch has still not been fully understood. The H1 and H4 receptor appear to be key components in the induction of itch. The
Arnold Bahlmann et al.
Chemosphere, 89(11), 1278-1286 (2012-06-12)
Carbamazepine (CBZ), caffeine and cetirizine were monitored by enzyme-linked immunosorbent assays (ELISAs) in surface and wastewaters from Berlin, Germany. This fast and cost-efficient method enabled to assess the spatial and temporal variation of these anthropogenic markers in a high-throughput screening.
Ko-Haung Lue et al.
International journal of pediatric otorhinolaryngology, 76(7), 994-1001 (2012-04-20)
Supplementary consumption of probiotics may temporarily alter the intestinal microflora of infants and children, thereby preventing and treating allergic disorders. To compare the clinical efficacy of levocetirizine with that of levocetirizine plus Lactobacillus johnsonii EM1 (Lj EM1) for treating perennial
Natalie J Carter
Drugs, 72(9), 1257-1269 (2012-06-13)
Bilastine is an orally administered, second-generation antihistamine used in the symptomatic treatment of seasonal or perennial allergic rhinoconjunctivitis and urticaria. In two well designed phase III trials, 14 days' treatment with bilastine was associated with a significantly lower area under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service