Skip to Content
Merck
All Photos(1)

Key Documents

89064

Sigma-Aldrich

4-Phenylazobenzyloxycarbonyl-Pro-Leu-Gly-Pro-D-Arg trifluoroacetate salt

collagenase substrate, chromogenic, ≥95% (HPLC), powder

Synonym(s):

Pz-Pro-Leu-Gly-Pro-D-Arg trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$259.00
250 MG
$585.00
1 G
$1,740.00

$259.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
100 MG
$259.00
250 MG
$585.00
1 G
$1,740.00

About This Item

Empirical Formula (Hill Notation):
C38H52N10O8 · xC2HF3O2
Molecular Weight:
776.88 (free base basis)
UNSPSC Code:
12352204
NACRES:
NA.32

$259.00


Estimated to ship onMay 29, 2025


Product Name

4-Phenylazobenzyloxycarbonyl-Pro-Leu-Gly-Pro-D-Arg trifluoroacetate salt, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

powder

Related Categories

Application

4-Phenylazobenzyloxycarbonyl-Pro-Leu-Gly-Pro-D-Arg trifluoroacetate salt has been used as substrate for collagenase.[1][2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Chikuma et al.
Journal of chromatography, 635(1), 81-87 (1993-04-09)
A rapid and sensitive assay method for the determination of PZ-peptidase activity is reported. This method is based on the monitoring of the absorption at 320 nm of 4-phenylazobenzyloxycarbonyl-L-Pro-L-Leu (PZ-Pro-Leu), enzymatically formed from the substrate 4-phenylazobenzyloxycarbonyl-L-Pro-L-Leu-Gly-L-Pro-D-Arg (PZ-peptide), after separation by
Highly sensitive assay for PZ-peptidase activity by high-performance liquid chromatography
Chikuma, T., et al.
Journal of Chromatography A, 348, 205-212 (1985)
M Nagelschmidt et al.
Biochimica et biophysica acta, 571(1), 105-111 (1979-11-09)
A peptidase cleaving a synthetic substrate for collagen peptidases, 4-phenylazobenzyloxcarbonyl-L-Pro-L-Leu-Gly-L-pro-D-Arg (designated as PZ-peptide) has been purified 1200-fold from rabbit serum and characterized. The enzyme preparation is free of collagenase and unspecific proteinase activity. The natural substrates are denatured collagen and
Collagenase H Is Crucial for Isolation of Rat Pancreatic Islets
Atsushi Fujio
Cell Transplantation (2014)
The induction of collagenase and a neutral proteinase by their high molecular weight substrates in Achromobacter iophagus.
V Keil-Dlouha et al.
Journal of molecular biology, 107(3), 293-305 (1976-11-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service