Skip to Content
Merck
All Photos(1)

Key Documents

85990

Sigma-Aldrich

Succinyl-β-cyclodextrin

Synonym(s):

succ-β-CD, succinyl-β-CD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

impurities

~5% water

color

white

mp

225  °C ((437 °F ) - Decomposes on heating)

storage temp.

−20°C

SMILES string

CC(=O)CCC(=O)OC[C@H]1O[C@@H]2O[C@H]3[C@@H](O)[C@H](O)[C@H](O[C@@H]3COC(=O)CCC(O)=O)O[C@H]4[C@@H](O)[C@H](O)[C@H](O[C@@H]4COC(=O)CCC(O)=O)O[C@H]5[C@@H](O)[C@H](O)[C@H](O[C@@H]5COC(=O)CCC(O)=O)O[C@H]6[C@@H](O)[C@H](O)[C@H](O[C@@H]6COC(=O)CCC(O)=O)O[C@H]7[C@@H](O)[C@H](O)[C@H](O[C@@H]7COC(=O)CCC(O)=O)O[C@H]8[C@@H](O)[C@H](O)[C@H](O[C@@H]8COC(=O)CCC(O)=O)O[C@H]1[C@@H](O)[C@@H]2O

InChI

1S/C71H100O55/c1-23(72)2-9-37(85)106-16-24-58-44(92)51(99)65(113-24)121-59-25(17-107-38(86)10-3-31(73)74)115-67(53(101)46(59)94)123-61-27(19-109-40(88)12-5-33(77)78)117-69(55(103)48(61)96)125-63-29(21-111-42(90)14-7-35(81)82)119-71(57(105)50(63)98)126-64-30(22-112-43(91)15-8-36(83)84)118-70(56(104)49(64)97)124-62-28(20-110-41(89)13-6-34(79)80)116-68(54(102)47(62)95)122-60-26(18-108-39(87)11-4-32(75)76)114-66(120-58)52(100)45(60)93/h24-30,44-71,92-105H,2-22H2,1H3,(H,73,74)(H,75,76)(H,77,78)(H,79,80)(H,81,82)(H,83,84)/t24-,25-,26-,27-,28-,29-,30-,44+,45+,46+,47+,48+,49+,50+,51+,52+,53+,54+,55+,56+,57+,58-,59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,71-/m1/s1

InChI key

DIRLEDPEXJLCIL-JCWBWLHSSA-N

General description

Succinyl-β-cyclodextrin is a negatively charged cyclodextrin derivative that has major implications in the chiral separation of neutral and basic analytes by means of capillary electrophoresis (CE), owing to its counter-current mobility.[1][2][3]

Application

Cyclodextrins (CD) are cyclic oligosaccharides composed of D-glucose units connected via α(1→4) glycosidic bonds. Common cyclodextrins contain six (α-CD); seven (β-CD) or eight (gamma-CD) D-glucose units. Cyclodextrins are sometimes derivitized through esterification at positions two, three and/or six.
Succinyl-β-cyclodextrin and carboxymethyl-β-cyclodextrin are used as chiral selective agents in capillary electrophoresis for the separation of di- and tri-peptide enantiomers and catechin enantiomers. Succinyl-β-cyclodextrin is used to optimize analysis of PNA-DNA duplexes with diethylthiadicarbocyanine dye.
Succinyl-β-cyclodextrin may be used as a chiral selector to resolve:
  • Acylamine fungicides in commercial agrochemical formulations by electrokinetic chromatography (EKC).[4][5]
  • Catechin isomers[6] in human biological samples and antihistamines[2] in pharmaceutical preparations by CE.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Sabbah et al.
Electrophoresis, 22(7), 1385-1393 (2001-05-31)
The separation of the LL and DD enantiomers of dipeptides and tripeptides using cyclodextrins (CDs) containing carboxyl groups was investigated with respect to the amino acid sequence of the peptides, the nature of the cyclodextrin and the buffer pH. Compared
Chiral Separations: Methods and Protocols
Methods in Molecular Biology (2004)
Influence of methanol on the enantioresolution of antihistamines with carboxymethyl-beta-cyclodextrin in capillary electrophoresis
Van Eeckhaut A, et al.
Electrophoresis, 25(16), 2838-2847 (2004)
Deia Abd el-Hady et al.
Talanta, 76(1), 138-145 (2008-07-01)
A simple and reliable analytical electrophoretic method using chiral capillary electrophoresis (CCE) with a high-sensitivity cell of special design has been established for simultaneous determination of (+)-catechin (C) and (-)-epicatechin (EC) in aqueous and human plasma media. The application of
Chiral separation of agricultural fungicides
Perez-Fernandez V, et al.
Journal of Chromatography A, 1218(38), 6561-6582 (2011)

Questions

  1. What is the molecular weight of this product?

    1 answer
    1. The molecular weight for the β-Cyclodextrin moiety is 1135 g/mole and the contribution from the succinic acid portion is 100.1 g/mole. This mass is used for succinic acid as water is lost during the condensation reaction when cyclodextrin and succinic acid form the conjugate. Thus, the average molecular formula and average formula weight can be expressed as the following. C42H70-nO35 • (C4H5O3)n or 1135.0 + n •100.1 The average degree of substitution for this product is ~3.5 for this product making the molecular weight approximately ~1485 g/mole for this product. The degree of substitution can vary from 2.5 - 5 for this product and the batch specific certificate of analysis should be reviewed to gather the degree of substitution (n).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service