Skip to Content
Merck
All Photos(1)

Key Documents

85278

Sigma-Aldrich

Mobile phase B for separation of amino acids on ABI® sequencers

Synonym(s):

Acetonitrile – 2-propanol mixture

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$191.00
2.5 L
$422.00

$191.00


Estimated to ship onMay 26, 2025



Select a Size

Change View
1 L
$191.00
2.5 L
$422.00

About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

$191.00


Estimated to ship onMay 26, 2025


form

liquid

density

0.78 g/mL at 20 °C

suitability

in accordance for sequence analysis test

storage temp.

2-8°C

SMILES string

CC#N.CC(C)O

InChI

1S/C3H8O.C2H3N/c1-3(2)4;1-2-3/h3-4H,1-2H3;1H3

InChI key

FPWNQPQTICPCOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Components

contains ~12% 2-propanol and ~88% acetonitrile

Legal Information

ABI PRISM is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service