83866
D-Ribose 1-phosphate bis(cyclohexylammonium) salt
≥99.0% (TLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H11O8P · 2C6H13N
CAS Number:
Molecular Weight:
428.46
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥99.0% (TLC)
form
powder
color
beige
storage temp.
−20°C
SMILES string
NC1CCCCC1.NC2CCCCC2.OC[C@H]3OC(OP(O)(O)=O)[C@H](O)[C@@H]3O
InChI
1S/2C6H13N.C5H11O8P/c2*7-6-4-2-1-3-5-6;6-1-2-3(7)4(8)5(12-2)13-14(9,10)11/h2*6H,1-5,7H2;2-8H,1H2,(H2,9,10,11)/t;;2-,3-,4-,5?/m..1/s1
InChI key
SSHMAENVNUVVFI-VNISEONSSA-N
Application
D-Ribose 1-phosphate is used to identify, differentiate and characterize phosphopentomutase(s) and nucleoside phosphorylases such as the uridine phosphorylase(s).
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service