Skip to Content
Merck
All Photos(1)

Key Documents

76399

Sigma-Aldrich

Ammonium sulfate 2M solution

Synonym(s):

Crystallization solution 32/Kit-No 82009

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$84.40
100 ML
$263.00

$84.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$84.40
100 ML
$263.00

About This Item

UNSPSC Code:
12352314
NACRES:
NA.26

$84.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

pH

6.0±0.2 ( in neat)

density

1.13 g/mL at 20 °C

storage temp.

2-8°C

Application


  • Optimization of nutrient-rich solution for direct fertigation using a novel side stream anaerobic forward osmosis process to treat textile wastewater.: This research optimizes nutrient-rich solutions for direct fertigation, highlighting the application of ammonium sulfate in the treatment and reuse of textile wastewater (Abbasi et al., 2021).

  • Electro-sorption of ammonium ion onto nickel foam supported highly microporous activated carbon prepared from agricultural residues (dried Luffa cylindrica).: This research explores the electro-sorption of ammonium ions using innovative materials, with ammonium sulfate solution facilitating the preparation process (Shih et al., 2019).

  • Structure of glycerol dehydrogenase (GldA) from Escherichia coli.: This structural biology study involves the use of ammonium sulfate in the crystallization process to elucidate the structure of an important enzyme from E. coli (Zhang et al., 2019).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service