Skip to Content
Merck

Skip To

75641

Poly(2′-deoxyguanyl-2′-deoxycytidylic acid) sodium salt

ampule of >10 A260

Synonym(s):

Poly(dG-dC) sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 U
Check Cart for Availability
$1,740.00

About This Item

CAS Number:
UNSPSC Code:
12352106
NACRES:
NA.52
MDL number:
Form:
liquid

$1,740.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

liquid

packaging

ampule of >10 A260

solubility

H2O: complete 1 mL/vial, clear, colorless

storage temp.

−20°C

SMILES string

[P](=O)(O[C@@H]3[C@H](O[C@H](C3)[n]4c5nc(nc(c5nc4)O)N)CO[P](=O)(O)O)(OC[C@H]1O[C@H](C[C@@H]1O)N2C=CC(=NC2=O)N)O

InChI

1S/C19H26N8O13P2/c20-12-1-2-26(19(30)23-12)13-3-8(28)10(38-13)5-37-42(34,35)40-9-4-14(39-11(9)6-36-41(31,32)33)27-7-22-15-16(27)24-18(21)25-17(15)29/h1-2,7-11,13-14,28H,3-6H2,(H,34,35)(H2,20,23,30)(H2,31,32,33)(H3,21,24,25,29)/t8-,9-,10+,11+,13+,14+/m0/s1

InChI key

IBZPKQZTZXRCKY-WKSZEZMPSA-N

Application

Poly (2′-deoxyguanyl-2′-deoxycytidylic acid) sodium salt was used as standard in gel electrophoresis to analyze the products of Klenow exo fragment of polymerase I.[1]

Biochem/physiol Actions

Poly (2′-deoxyguanyl-2′-deoxycytidylic acid) is a protonated polynucleotide structure frequently used to study the conformational changes of DNA molecules.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4929P938981349
form

liquid

form

powder

form

powder or solid

form

powder

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

H2O: complete 1 mL/vial, clear, colorless

solubility

-

solubility

-

solubility

water: 1 mL/vial, clear, colorless

packaging

ampule of >10 A260

packaging

-

packaging

-

packaging

ampule of ≥10 units A260

grade

Molecular Biology

grade

-

grade

-

grade

Molecular Biology


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters