Skip to Content
Merck

Skip To

75375

Ononin

≥99.0% (TLC)

Synonym(s):

Formononetin 7-O-β-D-glucopyranoside

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H22O9
CAS Number:
Molecular Weight:
430.40
UNSPSC Code:
12352201
NACRES:
NA.77
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
63067


assay

≥99.0% (TLC)

form

powder

SMILES string

COc1ccc(cc1)C2=COc3cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)ccc3C2=O

InChI

1S/C22H22O9/c1-28-12-4-2-11(3-5-12)15-10-29-16-8-13(6-7-14(16)18(15)24)30-22-21(27)20(26)19(25)17(9-23)31-22/h2-8,10,17,19-23,25-27H,9H2,1H3/t17-,19-,20+,21-,22-/m1/s1

InChI key

MGJLSBDCWOSMHL-MIUGBVLSSA-N

Gene Information

mouse ... Maoa(17161)
rat ... Aldh1a2(116676)

Biochem/physiol Actions

A isoflavone glucoside found in soybean, kudzu, and the Chinese medicinal herb, Radix astragali (RA). RA is a supplement taken to improve "energy." It may act by inducing erythropoietin expression.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Still not finding the right product?

Explore all of our products under Ononin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters