Skip to Content
Merck
All Photos(1)

Key Documents

73681

Sigma-Aldrich

4-Nitrophenyl α-D-maltohexaoside

≥95.0% (HPLC), powder

Synonym(s):

4-Nitrophenyl α-D-hexa-(1→4)-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$254.00
500 MG
$870.00

$254.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details



Select a Size

Change View
100 MG
$254.00
500 MG
$870.00

About This Item

Empirical Formula (Hill Notation):
C42H65NO33
CAS Number:
Molecular Weight:
1111.95
Beilstein:
3586152
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$254.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Product Name

4-Nitrophenyl α-D-maltohexaoside, ≥95.0% (HPLC)

Assay

≥95.0% (HPLC)

form

powder

impurities

≤5% water

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)Oc7ccc(cc7)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H65NO33/c44-5-13-19(50)20(51)26(57)38(66-13)72-33-15(7-46)68-40(28(59)22(33)53)74-35-17(9-48)70-42(30(61)24(35)55)76-36-18(10-49)71-41(31(62)25(36)56)75-34-16(8-47)69-39(29(60)23(34)54)73-32-14(6-45)67-37(27(58)21(32)52)65-12-3-1-11(2-4-12)43(63)64/h1-4,13-42,44-62H,5-10H2/t13-,14-,15-,16-,17-,18-,19-,20+,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37+,38-,39-,40-,41-,42-/m1/s1

InChI key

GNFSLPRCTKUSTG-UYDBUFRNSA-N

Application

4-Nitrophenyl ǥ-D-maltohexaoside has been used to assay ǥ-amylase enzyme activity.[1][2]

Biochem/physiol Actions

4-Nitrophenyl ǥ-D-maltohexaoside serves as a chromogenic substrate for ǥ-amylases and yields a yellow product upon cleavage[1] that can be spectrophotometrically measured at 44 nm.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Lorentz
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 21(7), 463-471 (1983-07-01)
Measurements of alpha-amylase with 4-nitrophenyl glucosides offer the following advantages over methods that rely on the formation of NADH: a short lag phase, no apparent interference by metabolites and enzymes of the sample and extremely stable substrates with low blank
Daniel P Birnbaum et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(11), 2004699-2004699 (2021-06-19)
Bacterial cellulose (BC) has excellent material properties and can be produced sustainably through simple bacterial culture, but BC-producing bacteria lack the extensive genetic toolkits of model organisms such as Escherichia coli (E. coli). Here, a simple approach is reported for
D G Semaan et al.
Journal of ethnopharmacology, 203, 39-46 (2017-03-28)
Ethno-botanical information from diabetic patients in Cuba led to the identification of Allophylus cominia as a possible source of new drugs for the treatment of type 2 diabetes mellitus (T2-DM). Chemical characterization of the extracts from A. cominia was carried
Daniel P Birnbaum et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(11), 2004699-2004699 (2021-06-19)
Bacterial cellulose (BC) has excellent material properties and can be produced sustainably through simple bacterial culture, but BC-producing bacteria lack the extensive genetic toolkits of model organisms such as Escherichia coli (E. coli). Here, a simple approach is reported for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service