Skip to Content
Merck
All Photos(1)

Key Documents

69769

Sigma-Aldrich

Microperoxidase disodium salt from horse heart

≥85% (HPCE)

Synonym(s):

H-11-PT, Haeme undecapeptide, MP-11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H114FeN20Na2O21S2
CAS Number:
Molecular Weight:
1905.88
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

≥85% (HPCE)

form

powder

impurities

peroxidase activity
≤10% water

solubility

H2O: 10 mg/mL, clear, dark red-brown

storage temp.

−20°C

SMILES string

O.O.O.O.[Na+].[Na+].CC(C)[C@H](N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@H]1CSSC(C)c2c(C)c3nc2nc4c(C)c(CCC([O-])=O)c5nc6nc(nc7c(C)c(C(C)SSC[C@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC1=O)C(=O)N[C@@H](Cc8c[nH]cn8)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(O)=O)C(O)=O)c(n3)n7[Fe]n45)c(C)c6CCC([O-])=O

InChI

1S/C80H113N23O22S4.Fe.2Na.4H2O/c1-32(2)59(83)77(121)89-47(19-24-55(110)111)72(116)88-45(15-13-14-26-81)71(115)92-50-29-126-128-41(12)58-37(8)65-101-68(58)99-63-35(6)44(17-23-54(108)109)67(98-63)103-66-43(16-22-53(106)107)34(5)62(97-66)96-64-36(7)57(69(100-64)102-65)40(11)129-127-30-51(93-73(117)46(18-21-52(82)105)87-70(114)38(9)86-75(50)119)76(120)91-49(27-42-28-84-31-85-42)74(118)95-61(39(10)104)79(123)94-60(33(3)4)78(122)90-48(80(124)125)20-25-56(112)113;;;;;;;/h28,31-33,38-41,45-51,59-61,104H,13-27,29-30,81,83H2,1-12H3,(H20,82,84,85,86,87,88,89,90,91,92,93,94,95,96,97,98,99,100,101,102,103,105,106,107,108,109,110,111,112,113,114,115,116,117,118,119,120,121,122,123,124,125);;;;4*1H2/q;+2;2*+1;;;;/p-4/t38-,39+,40?,41?,45-,46-,47-,48-,49-,50-,51-,59-,60-,61-;;;;;;;/m0......./s1

InChI key

HARTZTRJJQAYAK-FJHAOVHCSA-J

Antibody Adsorbant

Val-Glu-Lys-(Cys-Ala-Gln-Cys)Heme-His-Thr-Val-Glu

Preparation Note

prepared by enzymatic degradation of horse heart cytochrome c

Other Notes

Cationic heme undecapeptide as stain for detecting glycosaminoglycans on cellulose acetate; Kinetic study of pH-dependent properties
Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Wilson et al.
European journal of biochemistry, 77(1), 193-199 (1977-07-01)
The ferric form of the haem undecapeptide, derived from horse cytochrome c by peptic digestion, undergoes at least three pH-induced transitions with pK values of 3.4, 5.8 and 7.6. Temperature-jump experiments suggest that the first of these is due to
N Ghinea
Analytical biochemistry, 155(1), 78-82 (1986-05-15)
A rapid and sensitive method has been developed for detecting glycosaminoglycans on cellulose acetate strips. The method is based on the electrostatic interactions between glycosaminoglycans and cationic heme undecapeptide. The complexes are identified by peroxidatic reaction using 3,3'-diaminobenzidine as H2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service