Skip to Content
Merck

66540468

EDi017-C

human dermis (fibroblast)

Synonym(s):

EBiSC iPSC Line, Human iPSC, Induced Pluripotent Stem Cell Line, iPS Cell, iPSC

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352207
Biological source:
human dermis (fibroblast)
Relevant disease(s):
none
Growth mode:
adherent (pluripotent)
Manufacturer/tradename:
EBiSC

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

EDi017-C, Human iPS Cell Line

biological source

human dermis (fibroblast)

reprogramming method

Sendai virus

description

age (N/A)

manufacturer/tradename

EBiSC

gender

female

growth mode

adherent (pluripotent)

relevant disease(s)

none

shipped in

dry ice

storage temp.

−196°C

Quality Level

General description

Induced pluripotent stem cells (iPSCs) are adult cells that have been reprogrammed to an embryonic stem cell–like state. The cells can replicate indefinitely or, under controlled conditions, can be differentiated into any other cell type such as nerve, heart or liver cells. Medical researchers are able to use iPS cells to test how different patients might respond to new drugs or to analyse how genetic diseases develop.

The EBiSC stem cell bank is a collection of human iPS cells available to academic and commercial researchers for use in disease modelling and other forms of stem cell research. The initial collection has been generated from a wide range of donors representing specific disease backgrounds and healthy controls. EBiSC has established many routine procedures for collecting, expanding and characterizing human iPS cell lines. The stem cell bank includes iPSC cell lines derived from neurodegenerative diseases (Alzheimer′s Disease, Parkinson′s Disease, Dementia, Motor Neuron Disease (ALS) - and Huntington′s Disease), eye and heart diseases, and lines from healthy control donors for age and sex matching.

Biochem/physiol Actions

Depositor
University of Edinburgh
Derivation
Primary cell type: Fibroblast of dermis
Reprogramming method
Vector type: Non-integrating
Vector: Sendai virus
Gene list:
KLF4
MYC
POU5F1
SOX2
Is the reprogramming vector detectable: Unknown
other:From the same donor: EDi017-A, EDi017-B

Characterization
Analysis of Undifferentiated Cells
Marker expression: SSEA-1(-)TRA 1-60(+)POU5F1 (OCT-4)(+)SSEA-4(+)

Microbiology / Virus Screening
HIV 1: Negative
HIV 2: Not done
Hepatitis B: Negative
Hepatitis C: Negative
Mycoplasma: -

Genotyping
Karyotyping
Passage number: 22
Cell line karyotype: No autosomal or sex chromosome aneuploidies detected
Karyotyping method: BACs-on-Beads

Preparation Note

Surface coating: Matrigel®/Geltrex®
Passage method: Enzyme-free cell dissociation / EDTA
O2 concentration: 20%
CO2 concentration: 5%
Culture medium: Essential E8®

Other Notes

Note: EAUA and CLIP must be completed before order fulfillment

Legal Information

E8 is a registered trademark of WIsconsin Alumni Research Foundation non-stock Corporation
EBiSC is a trademark of Fraunhofer-Gesellschaft
GELTREX is a registered trademark of Life Technologies Corporation
Matrigel is a registered trademark of Corning, Inc.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service