Skip to Content
Merck
All Photos(2)

Key Documents

64054W

Midas® Vessels and Lids

new

Reagent containers for Midas® III-Plus Automated Stainer

Synonym(s):

Stainer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105202

shipped in

ambient

General description

Reagent containers for MIDAS® Automated Stainer

Components

Vessels and Lids to hold stain - 300 mL fill.

See-thru fill marks 50 mL, 75 mL, and 300 mL

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service