Skip to Content
Merck
All Photos(1)

Key Documents

59415C

SAFC

Phenol Red Solution 0.5%

suitable for cell culture

Synonym(s):

Phenol red solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

description

5.0 g/L phenol red
for research or for further manufacturing use

sterility

sterile; sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: 5.0 g/L

shipped in

ambient

SMILES string

O=S1(C2=CC=CC=C2C(C3=CC=C(O)C=C3)(C4=CC=C(O)C=C4)O1)=O

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared in Dulbecco′s Phosphate Buffered Saline.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisca Alcaraz-Pérez et al.
BMC biotechnology, 8, 81-81 (2008-10-29)
The dual-luciferase assay has been widely used in cell lines to determine rapidly but accurately the activity of a given promoter. Although this strategy has proved very useful, it does not allow the promoter and gene function to be analyzed
Pankaj Dixit et al.
Journal of pharmacological and toxicological methods, 65(1), 13-17 (2011-11-24)
Everted gut sac of rat intestine is a paradigm widely employed for determination of absorption kinetics of drugs along with evaluation of effects of absorption enhancers. Since its inception in 1954, it has been optimized to enhance tissue survival and
Diego Moldes et al.
TheScientificWorldJournal, 2012, 398725-398725 (2012-05-09)
The studies regarding decolorization of dyes by laccase may not only inform about the possible application of this enzyme for environmental purposes, but also may provide important information about its reaction mechanism and the influence of several factors that could
Adolf Michael Sandbichler et al.
Journal of comparative physiology. B, Biochemical, systemic, and environmental physiology, 181(8), 1023-1034 (2011-05-26)
A primary culture system of rainbow trout gill pavement cells grown on permeable support (single-seeded insert, SSI) was used to examine histological and physiological changes induced by the addition of the corticosteroid hormone cortisol. Pavement cell epithelia were cultured under
Héctor Rubén Hernández-Garcés et al.
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva, 104(1), 4-9 (2012-02-04)
An analytic study to validate a diagnostic test was carried out at the Institute of Gastroenterology in Havana, Cuba in adult patients of both sexes in whom chromoendoscopy was carried out with red phenol at 0.1% over the gastric mucosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service