Skip to Content
Merck
All Photos(1)

Key Documents

53657

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl nonanoate

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21BrClNO2
CAS Number:
Molecular Weight:
386.71
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98.0% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CCCCCCCCC(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12

InChI

1S/C17H21BrClNO2/c1-2-3-4-5-6-7-8-15(21)22-14-11-20-13-10-9-12(18)17(19)16(13)14/h9-11,20H,2-8H2,1H3

InChI key

WLSCZNTXZOTUFQ-UHFFFAOYSA-N

Application

suitable as substrate for lipase

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service