Skip to Content
Merck
All Photos(1)

Key Documents

53180

Sigma-Aldrich

Hexyl β-D-glucopyranoside

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$761.00

$761.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$761.00

About This Item

Empirical Formula (Hill Notation):
C12H24O6
CAS Number:
Molecular Weight:
264.32
Beilstein:
83239
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

$761.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

non-ionic

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]/D −36 to −31°° in H2O

mol wt

264.32 g/mol

technique(s)

RNA extraction: suitable
hybridization: suitable
tissue culture: suitable

CMC

250 mM

SMILES string

CCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H24O6/c1-2-3-4-5-6-17-12-11(16)10(15)9(14)8(7-13)18-12/h8-16H,2-7H2,1H3/t8-,9-,10+,11-,12-/m1/s1

InChI key

JVAZJLFFSJARQM-RMPHRYRLSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Non-ionic detergent used for solubilizing membrane proteins[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

n-Alkyl-glucosides as detergents for the preparation of highly homogeneous bilayer liposomes of variable sizes ( 60-240 nm phi) applying defined rates of detergent removal by dialysis.
R A Schwendener et al.
Biochemical and biophysical research communications, 100(3), 1055-1062 (1981-06-16)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service