51275
Hematoxylin solution according to Mayer
Synonym(s):
Mayer’s acid hemalum, Mordanted hematoxylin (Mayer)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
microbe id | staining: suitable
Quality Level
impurities
chloral hydrate
density
1.044 g/mL at 20 °C
Other Notes
Selective staining with hematoxylin, review; For staining nuclear material
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
PloS one, 8(8), e71641-e71641 (2013-08-24)
Fibroblast growth factor-4 (FGF4) is expressed in embryonic stages and in adult tissues, where it plays critical roles in modulating multiple cellular functions. However, the exact roles of FGF4 on proliferation and differentiation of embryonic stem cells (ESCs) are not
Frontiers in cell and developmental biology, 8, 597423-597423 (2021-01-08)
Recent studies reported that vascular endothelial cells (ECs) secrete NLR family pyrin domain-containing 3 (NLRP3) inflammasome products such as interleukin-1β (IL-1β) via extracellular vesicles (EVs) under various pathological conditions. EVs represent one of the critical mechanisms mediating the cell-to-cell communication
Conn?s Biological Stains, 345-345 (1969)
Conn?s Biological Stains, 8th edit., 345-345 (1969)
Scientific reports, 6, 21668-21668 (2016-02-27)
Externally visible body and longitudinal bone growth is a result of proliferation of chondrocytes. In growth disorder, there is delay in the age associated increase in height. The present study evaluates the effect of extract from transgenic tomato fruit expressing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service