506-05A
Human Lung Fibroblasts: HLF, adult
Synonym(s):
HLF, HLF cells
About This Item
Recommended Products
biological source
human lung (normal parenchyma)
Quality Level
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
karyotype
2n = 46
morphology
fibroblast
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
asthma; tuberculosis; cardiovascular diseases; cancer
shipped in
dry ice
storage temp.
−196°C
General description
HLF provide an excellent model system to study many aspects of human pulmonary function and pathophysiology.
HLF have been utilized in a number of research publications, for example to:
- Demonstrate differential gene expression patterns between fetal and adult lung fibroblasts, in particular showing that fetal fibroblasts express higher level of TGF-β receptor interacting protein 1 (TRIP-1) and consequently are less able to contract collagen than adult lung fibroblasts, leading to better healing of acute injuries in very immature lungs compared to higher levels of fibrosis observed in older patients (Navarro, 2009), and additionally discover that TRIP-1 regulates lung fibroblast/myofibroblast trans-differentiation (Nyp, 2014)
- Show that angiotensin II promotes lung fibrosis by activating expression of IL-4, IL-7, and PDGF-D increasing the extracellular matrix formation, and that this process can be blocked by angiotensin receptor inhibitor candesartan (Okada, 2009)
- Determine that phosphorylation of C/EBPβ contributes to lung fibrosis (Buck, 2011)
- Demonstrate that increased tissue rigidity activates toll-like receptor 2 and NF-κB in lung fibroblasts and induces expression of cytokines, CXCL1-3, IL-8 and TNF-α (Zheng, 2011)
- Demonstrate that 2-ethoxyestradiol, a non-estrogenic estradiol metabolite analog, inhibits cell growth and retards the progression of pulmonary fibrosis and hypertension (Tofovic, 2008, 2009)
- Identify the hypoxia-induced ROS/RhoA–ROCK dependent actin reorganization as a mediator of myofibroblast transformation and collagen synthesis in lung fibrosis (Ni, 2013)
Cell Line Origin
Application
Components
Preparation Note
- 1st passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
- Can be cultured at least 12 doublings
Subculture Routine
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service