Skip to Content
Merck
All Photos(1)

Key Documents

50008

Sigma-Aldrich

4-(2-Carboxyphenyl)-7-diethylamino-2-(4-dimethylaminophenyl)chromylium perchlorate

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$172.00

$172.00


Estimated to ship onMay 12, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$172.00

About This Item

Empirical Formula (Hill Notation):
C28H29ClN2O7
CAS Number:
Molecular Weight:
540.99
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

$172.00


Estimated to ship onMay 12, 2025


Request a Bulk Order

product line

BioReagent

Quality Level

suitability

in accordance for fluorescence
suitable for fluorescence

SMILES string

[O-]Cl(=O)(=O)=O.CCN(CC)c1ccc2c(cc([o+]c2c1)-c3ccc(cc3)N(C)C)-c4ccccc4C(O)=O

InChI

1S/C28H28N2O3.ClHO4/c1-5-30(6-2)21-15-16-23-25(22-9-7-8-10-24(22)28(31)32)18-26(33-27(23)17-21)19-11-13-20(14-12-19)29(3)4;2-1(3,4)5/h7-18H,5-6H2,1-4H3;(H,2,3,4,5)

InChI key

IZSVNKSOQVLCDH-UHFFFAOYSA-N

Other Notes

Cyanine-type fluorescent and laser dyes[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Czerney et al.
J. Photochem. Photobiol., A, 89, 31-31 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service